Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-nitropyridine N-oxide, 97%

$254.85 - $254.85

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-nitropyridine N-oxide
CAS 31872-57-8
Molecular Formula C5H4N2O4
Formula Weight 156.10g/mol
MDL Number MFCD00234062
Beilstein 1370608
Synonym 3-Nitro-4-pyridinol N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2004506

 
alfa aesar™
L20045-06
5g Each for $254.85
Description & Specifications

Specifications

Melting Point ≈225°C
Quantity 5g
Chemical Name or Material 4-Hydroxy-3-nitropyridine N-oxide
CAS 31872-57-8
Assay 97%
Molecular Formula C5H4N2O4
Formula Weight 156.10g/mol
MDL Number MFCD00234062
Beilstein 1370608
Synonym 3-Nitro-4-pyridinol N-oxide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.