Please login to your online account to display your discounted pricing

4-Hydroxy-3-nitropyridine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-nitropyridine
CAS 5435-54-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H4N2O3
Formula Weight 140.1
MDL Number MFCD00128874
Synonym 3-Nitro-4-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379460010

  SDS
thermo fisher scientific
AC379460010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379460050

  SDS
thermo fisher scientific
AC379460050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 140.1
Quantity 1g
Color Light yellow
CAS Max % 100.0
Chemical Name or Material 4-Hydroxy-3-nitropyridine
Molecular Formula C5H4N2O3
MDL Number MFCD00128874
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 99%
CAS 5435-54-1
Synonym 3-Nitro-4-pyridinol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.