missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-3-nitrocoumarin 98.0+%, TCI America™

$128.38 - $444.39

Chemical Identifiers

CAS 20261-31-8
Molecular Formula C9H5NO5
Molecular Weight (g/mol) 207.141
MDL Number MFCD00051671
InChI Key NZQAQAUWFHMVEM-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
PubChem CID 54682640
IUPAC Name 4-hydroxy-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14585G View Documents Tci America
H14585G
5g Each for $128.38
 
H145825G View Documents Tci America
H145825G
25g Each for $444.39
 
Specifications

Chemical Identifiers

20261-31-8
207.141
NZQAQAUWFHMVEM-UHFFFAOYSA-N
54682640
C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O
C9H5NO5
MFCD00051671
4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
4-hydroxy-3-nitrochromen-2-one

Specifications

Yellow
5g
C9H5NO5
4-hydroxy-3-nitrocoumarin, 4-hydroxy-3-nitro-2h-chromen-2-one, 3-nitro-4-hydroxycoumarin, 2h-1-benzopyran-2-one, 4-hydroxy-3-nitro, 2-hydroxy-3-nitrochromen-4-one, 4-hydroxy-3-nitro-chromen-2-one, 2h-1-benzopyran-2-one,4-hydroxy-3-nitro, 4-hydroxy-3-hydroxy oxido amino-2h-chromen-2-one, acmc-20amh1, maybridge1_005221
C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])O
207.141
207.14
≥98.0% (HPLC,T)
171°C
20261-31-8
MFCD00051671
NZQAQAUWFHMVEM-UHFFFAOYSA-N
4-hydroxy-3-nitrochromen-2-one
54682640
Crystal-Powder at 20°C
4-Hydroxy-3-nitrocoumarin
Safety and Handling

Safety and Handling

TSCA : No