Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-nitrobiphenyl, 97%

$91.91 - $317.09

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-nitrobiphenyl
CAS 885-82-5
Molecular Formula C12H9NO3
Formula Weight 215.21
MDL Number MFCD00014694
Beilstein 2331623
Synonym 2-Nitro-4-phenylphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0277309

 
alfa aesar™
L0277309
10g Each for $91.91

AAL0277318

 
alfa aesar™
L0277318
50g Each for $317.09
Description & Specifications

Specifications

Formula Weight 215.21
Chemical Name or Material 4-Hydroxy-3-nitrobiphenyl
Assay 97%
Molecular Formula C12H9NO3
CAS 885-82-5
Synonym 2-Nitro-4-phenylphenol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 65° to 69°C
MDL Number MFCD00014694
Beilstein 2331623
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.