missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-nitrobenzonitrile, 98%

Manufacturer:  Alfa Aesar™ H5559703

 View more versions of this product

Catalog No. AAH5559703



Specifications

Specifications

4-Hydroxy-3-nitrobenzonitrile
1g
C7H4N2O3
2048724
INBLGVOPOSGVTA-UHFFFAOYSA-N
164.12
164.12
146°C to 148°C
3272-08-0
MFCD00070775
2-nitro-4-cyanophenol
C1=CC(=C(C=C1C#N)[N+](=O)[O-])O
76758
98%

Chemical Identifiers

3272-08-0
164.12
INBLGVOPOSGVTA-UHFFFAOYSA-N
76758
C7H4N2O3
MFCD00070775
2-nitro-4-cyanophenol
C1=CC(=C(C=C1C#N)[N+](=O)[O-])O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 221-899-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents