Please login to your online account to display your discounted pricing

4-Hydroxy-3-nitrobenzoic acid, 98%, ACROS Organics™

$131.35 - $131.35

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-nitrobenzoic acid
Name Note 98%
CAS 616-82-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H5NO5
Linear Formula HOC6H3(NO2)CO2H
Formula Weight 183.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC211521000

  SDS
thermo fisher scientific
211521000
Glass bottle 100g Each for $131.35
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 183.12
Physical Form Powder
Quantity 100g
Color Yellow to light brown
EINECS Number 210-494-1
Melting Point 182° to 186°C
Beilstein 10, 181
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
MDL Number MFCD00007119
Assay Percent Range 98%
Linear Formula HOC6H3(NO2)CO2H
Name Note 98%
Chemical Name or Material 4-Hydroxy-3-nitrobenzoic acid
CAS 616-82-0
Molecular Formula C7H5NO5
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.