Please login to your online account to display your discounted pricing

4-Hydroxy-3-nitrobenzenearsonic Acid 98%, ACROS Organics™

$20.37 - $20.37

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-nitrobenzenearsonic acid
Name Note 98%
CAS 121-19-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C6H6AsNO6
Linear Formula HOC6H3(NO2)AsO3H2
Formula Weight 263.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121660050

  SDS
thermo fisher scientific
AC121660050
Glass bottle 5g Each for $20.37
Description & Specifications

Specifications

Assay ≥97.5% (on dry substance)
Formula Weight 263.03
Physical Form Fine Crystalline Powder
Quantity 5g
Color Beige-brown
RTECS Number CY5250000
TSCA TSCA
EINECS Number 204-453-7
Beilstein 16, I, 456
Merck Index 15, 8406
CAS Max % 100.0
Water 1% max.
Synonym Nitrophenol-4-arsenic acid
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Molecular Formula C6H6AsNO6
MDL Number MFCD00007112
Chemical Name or Material 4-Hydroxy-3-nitrobenzenearsonic acid
CAS 121-19-7
Name Note 98%
Linear Formula HOC6H3(NO2)AsO3H2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.