missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxy-3'-nitroacetophenone, 98%

$42.96 - $42.96

Chemical Identifiers

CAS 6322-56-1
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00017002
Beilstein 1959078
Chemical Name or Material 4'-Hydroxy-3'-nitroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1831703 SDS Alfa Aesar™
A1831703
1g Each for $42.96
AAA1831706 SDS Alfa Aesar™
A1831706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1831714 SDS Alfa Aesar™
A1831714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6322-56-1
181.15
1959078
C8H7NO4
MFCD00017002
4'-Hydroxy-3'-nitroacetophenone

Specifications

131° to 133°C
6322-56-1
181.15
1959078
98%
1g
C8H7NO4
MFCD00017002
4'-Hydroxy-3'-nitroacetophenone

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning