Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-methylpyridine, 97%

$127.01 - $351.08

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-methylpyridine
CAS 22280-02-0
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD04114239
Synonym 4-Hydroxy-2-picoline; 3-Methyl-4-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH32265MD

  SDS
alfa aesar™
H32265MD
250mg Each for $127.01

AAH3226503

  SDS
alfa aesar™
H3226503
1g Each for $351.08
Description & Specifications

Specifications

Formula Weight 109.13
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 4-Hydroxy-3-methylpyridine
Synonym 4-Hydroxy-2-picoline; 3-Methyl-4-pyridinol
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 250mg
Melting Point ≈168°C
Molecular Formula C6H7NO
MDL Number MFCD04114239
CAS 22280-02-0

Applications
4-Hydroxy-3-methylpyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (14 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.