Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-methylbenzoic acid hydrate, 97%

$31.34 - $152.19

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-methylbenzoic acid hydrate
CAS 499-76-3
Molecular Formula C8H8O3·xH2O
Formula Weight 152.15 (Anhydrous)
MDL Number MFCD00270105
Synonym 4-Hydroxy-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2779203

  SDS
alfa aesar™
H2779203
1g Each for $31.34

AAH2779209

  SDS
alfa aesar™
H2779209
10g Each for $152.19
Description & Specifications

Specifications

Formula Weight 152.15 (Anhydrous)
Assay 97%
CAS 499-76-3
Quantity 1g
Molecular Formula C8H8O3·xH2O
Melting Point 173° to 177°C
Chemical Name or Material 4-Hydroxy-3-methylbenzoic acid hydrate
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD00270105
Synonym 4-Hydroxy-m-toluic acid
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.