Please login to your online account to display your discounted pricing

4'-Hydroxy-3'-methylacetophenone 99.5+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  121400250

 View more versions of this product

Catalog No. AC121400250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Formula Weight 150.18
Physical Form Crystalline Powder or Crystals
Quantity 25g
Color White to light beige
Beilstein 08, 112
TSCA TSCA
EINECS Number 212-880-5
Melting Point 107° to 109°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
CAS Min % 99.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 876-02-8
Linear Formula HOC6H3(CH3)COCH3
Assay Percent Range 99.5+%
MDL Number MFCD00002231
Molecular Formula C9 H10 O2
Name Note 99.5+%
Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone

Specifications

Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone
Name Note 99.5+%
CAS 876-02-8
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C9 H10 O2
Linear Formula HOC6H3(CH3)COCH3
Formula Weight 150.18
MDL Number MFCD00002231
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents