Please login to your online account to display your discounted pricing

4'-Hydroxy-3'-methylacetophenone 99.5+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone
Name Note 99.5+%
CAS 876-02-8
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C9 H10 O2
Linear Formula HOC6H3(CH3)COCH3
Formula Weight 150.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC121400250

  SDS
thermo fisher scientific
121400250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.50%
Formula Weight 150.18
Physical Form Crystalline Powder or Crystals
Quantity 25g
Color White to light beige
Beilstein 08, 112
EINECS Number 212-880-5
TSCA TSCA
Melting Point 107° to 109°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 4'-Hydroxy-3'-methylacetophenone
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99.5+%
Assay Percent Range 99.5+%
Molecular Formula C9 H10 O2
Linear Formula HOC6H3(CH3)COCH3
MDL Number MFCD00002231
CAS 876-02-8
CAS Min % 99.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.