Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-methoxyphenylacetone, 95%

$39.70 - $109.92

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-methoxyphenylacetone
CAS 2503-46-0
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00130260
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1972803

  SDS
alfa aesar™
A1972803
1g Each for $39.70

AAA1972806

  SDS
alfa aesar™
A1972806
5g Each for $109.92
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material 4-Hydroxy-3-methoxyphenylacetone
Assay 95%
Molecular Formula C10H12O3
MDL Number MFCD00130260
CAS 2503-46-0
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-propanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 126° to 127°C (0.3mmHg)
Quantity 1g

Applications
It is often found in wine that has been aged in wood barrels contributing to the wine’s flavor profile.

Solubility
Soluble in alcohol and water, 1.101e+004 mg/L @ 25°C (est.).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.