missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-3-methoxymandelic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 164900010

 View more versions of this product

Catalog No. AC164900010



Specifications

Specifications

4-Hydroxy-3-methoxymandelic acid
Glass bottle
131.0°C to 134.0°C
55-10-7
100.0
C9H10O5
MFCD00004235
+/--vanillylmandelic acid
CGQCWMIAEPEHNQ-UHFFFAOYSA-N
2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
1245
198.17
99%
Authentic
White to Beige
1g
98.5
99%
HOC6H3(OCH3)CH(OH)CO2H
15,10121
Solubility in water: freely soluble. Other solubilities: freely soluble in acetone,moderatly soluble in ether and acetonitrile,sparingly soluble in benzene
COC1=C(C=CC(=C1)C(C(=O)O)O)O
198.17
CHEBI:20106
Fine Crystalline Powder

Chemical Identifiers

55-10-7
198.17
CGQCWMIAEPEHNQ-UHFFFAOYSA-N
1245
2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
C9H10O5
MFCD00004235
+/--vanillylmandelic acid
CHEBI:20106
COC1=C(C=CC(=C1)C(C(=O)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-224-

SDS Documents