Learn More
4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156360050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Hydroxy-3-methoxycinnamic acid | |
1135-24-6 | |
100.0 | |
White to Beige | |
98.5% min. (HPLC) | |
C10H10O4 | |
MFCD00004400 | |
10, 436 | |
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
KSEBMYQBYZTDHS-HWKANZROSA-N | |
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid | |
445858 | |
194.19 | |
Powder |
99% | |
98.5 | |
169.0°C to 173.0°C | |
Authentic | |
Glass bottle | |
HOC6H3(OCH3)CH=CHCO2H | |
5 g | |
15, 4089 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ethyl acetate,moderately soluble in diethyl ether,sparingly soluble in petroleum ether and benzene | |
COC1=C(C=CC(=C1)C=CC(=O)O)O | |
194.19 | |
CHEBI:17620 | |
99% |
Chemical Identifiers
1135-24-6 | |
194.19 | |
KSEBMYQBYZTDHS-HWKANZROSA-N | |
445858 | |
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid |
C10H10O4 | |
MFCD00004400 | |
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
CHEBI:17620 | |
COC1=C(C=CC(=C1)C=CC(=O)O)O |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 214-490-0
RTECSNumber : UD3365500
TSCA : TSCA
RUO – Research Use Only