Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%

$113.01 - $113.01

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
CAS 7149-10-2
Molecular Formula C8H11NO2·HCl
Formula Weight 189.64
MDL Number MFCD00012864
Beilstein 3915418
Synonym Vanillylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2063706

  SDS
alfa aesar™
B2063706
5g Each for $113.01
Description & Specifications

Specifications

Formula Weight 189.64
Synonym Vanillylamine hydrochloride
Health Hazard 1 Warning
Beilstein 3915418
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
Molecular Formula C8H11NO2·HCl
CAS 7149-10-2
Quantity 5g
Melting Point ≈220°C (dec.)
MDL Number MFCD00012864
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.