missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 174582500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Hydroxy-3-methoxybenzyl alcohol | |
498-00-0 | |
100.0 | |
98.5% min. (GC) | |
C8H10O3 | |
MFCD00004659 | |
06,IV,7381 | |
ZENOXNGFMSCLLL-UHFFFAOYSA-N | |
4-(hydroxymethyl)-2-methoxyphenol | |
62348 | |
154.16 |
99% | |
98.5 | |
Authentic | |
Plastic bottle | |
HOC6H3(OCH3)CH2OH | |
250g | |
vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol | |
COC1=CC(CO)=CC=C1O | |
154.17 | |
CHEBI:18353 | |
99% |
Chemical Identifiers
498-00-0 | |
154.17 | |
ZENOXNGFMSCLLL-UHFFFAOYSA-N | |
62348 | |
4-(hydroxymethyl)-2-methoxyphenol |
C8H10O3 | |
MFCD00004659 | |
vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol | |
CHEBI:18353 | |
COC1=CC(CO)=CC=C1O |
Safety and Handling
EINECSNumber : 207-852-4
RUO â Research Use Only