Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3-methoxybenzyl alcohol, 98+%

$45.81 - $171.77

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol
CAS 498-00-0
Molecular Formula C8H10O3
Formula Weight 154.17
MDL Number MFCD00004659
Beilstein 1910044
Synonym Vanillyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1516030

  SDS
alfa aesar™
A1516030
250g Each for $171.77

AAA1516018

  SDS
alfa aesar™
A1516018
50g Each for $45.81
Description & Specifications

Specifications

Formula Weight 154.17
Beilstein 1910044
Melting Point 113° to 115°C
Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 498-00-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H10O3
Synonym Vanillyl alcohol
MDL Number MFCD00004659
Quantity 250g
Health Hazard 1 Warning
Assay ≥98%

Applications
Used as a flavoring agent. Other possible uses, vanilla, coconut, cream and other dairy nuances, coumarin.

Solubility
Soluble in 95% ethanol (5 %), water, and oils.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.