missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-3-methoxybenzoic acid, 97+%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 121350250

 View more versions of this product

Catalog No. AC121350250



Description

Description

Specifications

Specifications

4-Hydroxy-3-methoxybenzoic acid
Glass bottle
Brown to Colorless
25g
121-34-6
100.0
C8H8O4
MFCD00002551
15, 10119
Solubility in water: .. Other solubilities: soluble in alcohol and ether
COC1=CC(=CC=C1O)C(O)=O
168.15
CHEBI:30816
Liquid
Authentic
0.5% max. (K.F.)
210°C to 214°C
98.5+%
98.5
97% min. (HPLC)
HOC6H3(OCH3)CO2H
10, 392
3-methoxy-4-hydroxybenzoic acid, 4-hydroxy-3-methoxy-benzoic acid, acide vanillique, benzoic acid, 4-hydroxy-3-methoxy, m-anisic acid, 4-hydroxy, p-vanillic acid, protocatechuic acid, 3-methyl ester, unii-gm8q3jm2y8, vanillate, vanillic acid
WKOLLVMJNQIZCI-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzoic acid
8468
168.15
97+%

Chemical Identifiers

121-34-6
168.15
WKOLLVMJNQIZCI-UHFFFAOYSA-N
8468
4-hydroxy-3-methoxybenzoic acid
C8H8O4
MFCD00002551
3-methoxy-4-hydroxybenzoic acid, 4-hydroxy-3-methoxy-benzoic acid, acide vanillique, benzoic acid, 4-hydroxy-3-methoxy, m-anisic acid, 4-hydroxy, p-vanillic acid, protocatechuic acid, 3-methyl ester, unii-gm8q3jm2y8, vanillate, vanillic acid
CHEBI:30816
COC1=CC(=CC=C1O)C(O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-466-8

RTECSNumber : YW5300000

TSCA : TSCA

SDS Documents
Product Certifications