missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxy-3-methoxy-alpha-methylbenzyl Alcohol, TCI America™
$139.03 - $469.03
Chemical Identifiers
CAS | 2480-86-6 |
---|---|
Molecular Formula | C9H12O3 |
Molecular Weight (g/mol) | 168.192 |
MDL Number | MFCD00238638 |
InChI Key | BDRRAMWDUCXAKG-UHFFFAOYSA-N |
Synonym | 2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol |
PubChem CID | 17203 |
ChEBI | CHEBI:86590 |
IUPAC Name | 4-(1-hydroxyethyl)-2-methoxyphenol |
SMILES | CC(C1=CC(=C(C=C1)O)OC)O |
Chemical Identifiers
2480-86-6 | |
168.192 | |
BDRRAMWDUCXAKG-UHFFFAOYSA-N | |
17203 | |
4-(1-hydroxyethyl)-2-methoxyphenol |
C9H12O3 | |
MFCD00238638 | |
2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol | |
CHEBI:86590 | |
CC(C1=CC(=C(C=C1)O)OC)O |
Specifications
Undesignated | |
1g | |
C9H12O3 | |
2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol | |
CC(C1=CC(=C(C=C1)O)OC)O | |
168.192 | |
CHEBI:86590 | |
Crystal-Powder at 20°C | |
4-Hydroxy-3-methoxy-alpha-methylbenzyl Alcohol |
102°C | |
2480-86-6 | |
MFCD00238638 | |
BDRRAMWDUCXAKG-UHFFFAOYSA-N | |
4-(1-hydroxyethyl)-2-methoxyphenol | |
17203 | |
168.19 | |
≥98.0% (GC) |
Safety and Handling
TSCA : No