Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dinitrophenylacetic acid, 97%

$38.12 - $116.77

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3,5-dinitrophenylacetic acid
CAS 10463-37-3
Molecular Formula C8H6N2O7
Formula Weight 242.15
MDL Number MFCD00016995
Beilstein 2665595
Synonym 3,5-Dinitro-4-hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1164303

  SDS
alfa aesar™
L1164303
1g Each for $38.12

AAL1164306

  SDS
alfa aesar™
L1164306
5g Each for $116.77
Description & Specifications

Specifications

Formula Weight 242.15
CAS 10463-37-3
Chemical Name or Material 4-Hydroxy-3,5-dinitrophenylacetic acid
MDL Number MFCD00016995
Beilstein 2665595
Synonym 3,5-Dinitro-4-hydroxyphenylacetic acid
Assay 97%
Molecular Formula C8H6N2O7
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 166° to 176°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.