missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dinitrophenylacetic acid, 97%

Manufacturer:  Alfa Aesar™ L1164306

 View more versions of this product

Catalog No. AAL1164306



Specifications

Specifications

4-Hydroxy-3,5-dinitrophenylacetic acid
5g
C8H6N2O7
2665595
MLVYQQLUGFSXQH-UHFFFAOYSA-N
242.143
242.15
166°C to 176°C
10463-37-3
MFCD00016995
3,5-dinitro-4-hydroxyphenylacetic acid, 4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrophenylacetic acid, 3,5-dinitro-4-hydroxyphenacetic acid, 4-hydroxy-3,5-dinitro-phenyl-acetic acid, 2-4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrobenzeneacetic acid, 4-hydroxy-3,5-dinitrophenacetyl, acmc-20anc2, 3,5-dinitro-4-hydroxyphenyl-acetic acid
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(=O)O
98596
97%

Chemical Identifiers

10463-37-3
242.143
MLVYQQLUGFSXQH-UHFFFAOYSA-N
98596
C8H6N2O7
MFCD00016995
3,5-dinitro-4-hydroxyphenylacetic acid, 4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrophenylacetic acid, 3,5-dinitro-4-hydroxyphenacetic acid, 4-hydroxy-3,5-dinitro-phenyl-acetic acid, 2-4-hydroxy-3,5-dinitrophenyl acetic acid, 4-hydroxy-3,5-dinitrobenzeneacetic acid, 4-hydroxy-3,5-dinitrophenacetyl, acmc-20anc2, 3,5-dinitro-4-hydroxyphenyl-acetic acid
C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents