Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dinitrobenzoic acid, 98+%

$306.31 - $306.31

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-3,5-dinitrobenzoic acid
CAS 1019-52-9
Molecular Formula C7H4N2O7
Formula Weight 228.12
MDL Number MFCD00017000
Synonym 3,5-Dinitro-4-hydroxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2247406

 
alfa aesar™
B2247406
5g Each for $306.31
Description & Specifications

Specifications

Formula Weight 228.12
Chemical Name or Material 4-Hydroxy-3,5-dinitrobenzoic acid
Assay ≥98%
Molecular Formula C7H4N2O7
MDL Number MFCD00017000
CAS 1019-52-9
Synonym 3,5-Dinitro-4-hydroxybenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 248° to 252°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.