missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%

$39.70 - $421.76

Chemical Identifiers

CAS 530-56-3
Molecular Formula C9H12O4
Formula Weight 184.19
MDL Number MFCD00016871
Beilstein 1873975
Synonym 3,5-Dimethoxy-4-hydroxybenzyl alcohol; Syringic alcohol
Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzyl alcohol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1388803 SDS Alfa Aesar™
A1388803
1g Each for $39.70
AAA1388814 SDS Alfa Aesar™
A1388814
25g Each for $421.76
AAA1388806 SDS Alfa Aesar™
A1388806
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

Solubility
Sparingly soluble (0.40 g/L at 25°C).

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

530-56-3
184.19
1873975
4-Hydroxy-3,5-dimethoxybenzyl alcohol
C9H12O4
MFCD00016871
3,5-Dimethoxy-4-hydroxybenzyl alcohol; Syringic alcohol

Specifications

129° to 132°C
530-56-3
184.19
1873975
4-Hydroxy-3,5-dimethoxybenzyl alcohol
1g
C9H12O4
MFCD00016871
3,5-Dimethoxy-4-hydroxybenzyl alcohol; Syringic alcohol
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning