missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dimethoxybenzhydrazide, 97%

$63.88 - $222.89

Chemical Identifiers

CAS 1443-76-1
Molecular Formula C9H12N2O4
Formula Weight 212.21
MDL Number MFCD00017077
Beilstein 2657142
Synonym 3,5-Dimethoxy-4-hydroxybenzhydrazide
Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzhydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1103206 SDS Alfa Aesar™
L1103206
5g Each for $222.89
AAL1103203 SDS Alfa Aesar™
L1103203
1g Each for $63.88
Specifications Description & Specifications

Chemical Identifiers

1443-76-1
212.21
2657142
4-Hydroxy-3,5-dimethoxybenzhydrazide
C9H12N2O4
MFCD00017077
3,5-Dimethoxy-4-hydroxybenzhydrazide

Specifications

1443-76-1
154° to 157°C
MFCD00017077
5g
4-Hydroxy-3,5-dimethoxybenzhydrazide
C9H12N2O4
212.21
2657142
3,5-Dimethoxy-4-hydroxybenzhydrazide
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning