missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-3,5-dimethoxybenzhydrazide, 97%

Manufacturer:  Alfa Aesar™ L1103203

 View more versions of this product

Catalog No. AAL1103203



Specifications

Specifications

4-Hydroxy-3,5-dimethoxybenzhydrazide
1g
C9H12N2O4
2657142
AKZAAANGIPRVDU-UHFFFAOYSA-N
4-hydroxy-3,5-dimethoxybenzohydrazide
74043
97%
154°C to 157°C
1443-76-1
MFCD00017077
3,5-dimethoxy-4-hydroxybenzhydrazide, 4-hydroxy-3,5-dimethoxybenzhydrazide, benzoic acid, 4-hydroxy-3,5-dimethoxy-, hydrazide, acmc-1bzfx, 4-???-2,6-dimethoxyphenol, #, 4-hydroxy-3,5-dimethoxybenzhydrazide, 3,5-dimethoxy-4-hydroxy benzhydrazide, 4-hydroxy-3,5-dimethoxy-benzoic acid hydrazide, benzoic acid,4-hydroxy-3,5-dimethoxy-, hydrazide
COC1=CC(=CC(=C1O)OC)C(=O)NN
212.205
212.21

Chemical Identifiers

1443-76-1
212.205
AKZAAANGIPRVDU-UHFFFAOYSA-N
74043
COC1=CC(=CC(=C1O)OC)C(=O)NN
C9H12N2O4
MFCD00017077
3,5-dimethoxy-4-hydroxybenzhydrazide, 4-hydroxy-3,5-dimethoxybenzhydrazide, benzoic acid, 4-hydroxy-3,5-dimethoxy-, hydrazide, acmc-1bzfx, 4-???-2,6-dimethoxyphenol, #, 4-hydroxy-3,5-dimethoxybenzhydrazide, 3,5-dimethoxy-4-hydroxy benzhydrazide, 4-hydroxy-3,5-dimethoxy-benzoic acid hydrazide, benzoic acid,4-hydroxy-3,5-dimethoxy-, hydrazide
4-hydroxy-3,5-dimethoxybenzohydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents