Learn More
4-Hydroxy-3,5-diiodobenzoic acid, 97%, Thermo Scientific Chemicals
$136.12 - $547.03
Chemical Identifiers
CAS | 618-76-8 |
---|---|
Molecular Formula | C7H4I2O3 |
Molecular Weight (g/mol) | 389.92 |
MDL Number | MFCD00016532 |
InChI Key | XREKTVACBXQCSB-UHFFFAOYSA-N |
Synonym | 3,5-diiodo-4-hydroxybenzoic acid, ioxynil acid, ioxynic acid, benzoic acid, 4-hydroxy-3,5-diiodo, unii-8c0qv5jk0d, 4-hydroxy-3,5-diiodo-benzoic acid, 8c0qv5jk0d, chembl83650, benzoic acid, 3,5-diiodo-4-hydroxy, acmc-1b674 |
PubChem CID | 12065 |
IUPAC Name | 4-hydroxy-3,5-diiodobenzoic acid |
SMILES | OC(=O)C1=CC(I)=C(O)C(I)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0259509
|
Thermo Scientific Chemicals
L0259509 |
10 g |
Each for $136.12
|
|
|||||
AAL0259518
|
Thermo Scientific Chemicals
L0259518 |
50 g |
Each for $547.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
618-76-8 | |
389.92 | |
XREKTVACBXQCSB-UHFFFAOYSA-N | |
12065 | |
OC(=O)C1=CC(I)=C(O)C(I)=C1 |
C7H4I2O3 | |
MFCD00016532 | |
3,5-diiodo-4-hydroxybenzoic acid, ioxynil acid, ioxynic acid, benzoic acid, 4-hydroxy-3,5-diiodo, unii-8c0qv5jk0d, 4-hydroxy-3,5-diiodo-benzoic acid, 8c0qv5jk0d, chembl83650, benzoic acid, 3,5-diiodo-4-hydroxy, acmc-1b674 | |
4-hydroxy-3,5-diiodobenzoic acid |
Specifications
618-76-8 | |
C7H4I2O3 | |
10 g | |
Light sensitive | |
XREKTVACBXQCSB-UHFFFAOYSA-N | |
4-hydroxy-3,5-diiodobenzoic acid | |
12065 | |
97% |
∽275°C (decomposition) | |
MFCD00016532 | |
2212145 | |
3,5-diiodo-4-hydroxybenzoic acid, ioxynil acid, ioxynic acid, benzoic acid, 4-hydroxy-3,5-diiodo, unii-8c0qv5jk0d, 4-hydroxy-3,5-diiodo-benzoic acid, 8c0qv5jk0d, chembl83650, benzoic acid, 3,5-diiodo-4-hydroxy, acmc-1b674 | |
OC(=O)C1=CC(I)=C(O)C(I)=C1 | |
389.92 | |
389.91 | |
4-Hydroxy-3,5-diiodobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-562-0
RTECSNumber : DG8597500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only