Learn More
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 454680100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin | |
1% max. | |
96% min. (HPLC) | |
10g | |
warfarin, coumadin, coumafene, prothromadin, coumafen, coumarins, zoocoumarin, coumefene, panwarfin, warfarine | |
PJVWKTKQMONHTI-UHFFFAOYSA-N | |
4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one | |
54678486 | |
308.34 |
81-81-2 | |
Authentic | |
C19H16O4 | |
15,10236 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone, dioxane, moderatly soluble in, methanol, ethanol, isopropanol, insoluble in benzene, cyclohexane | |
CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O | |
308.34 | |
CHEBI:87732 | |
97% |
Chemical Identifiers
81-81-2 | |
308.34 | |
warfarin, coumadin, coumafene, prothromadin, coumafen, coumarins, zoocoumarin, coumefene, panwarfin, warfarine | |
CHEBI:87732 | |
CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O |
C19H16O4 | |
PJVWKTKQMONHTI-UHFFFAOYSA-N | |
54678486 | |
4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one |
Safety and Handling
GHS H Statement
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
GHS P Statement
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 201-377-6
RUO â Research Use Only