missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-2-(trifluoromethyl)pyridine, TCI America™

$105.42 - $368.20

Chemical Identifiers

CAS 170886-13-2
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.099
MDL Number MFCD01571202
InChI Key LKHCWMQMRXAPHX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
PubChem CID 10953849
IUPAC Name 2-(trifluoromethyl)-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15285G View Documents TCI America
H15285G
5g Each for $368.20
 
H15281G View Documents TCI America
H15281G
1g Each for $105.42
 
Specifications

Chemical Identifiers

170886-13-2
163.099
LKHCWMQMRXAPHX-UHFFFAOYSA-N
10953849
C1=CNC(=CC1=O)C(F)(F)F
C6H4F3NO
MFCD01571202
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
2-(trifluoromethyl)-1H-pyridin-4-one

Specifications

White-Yellow
5g
C6H4F3NO
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
C1=CNC(=CC1=O)C(F)(F)F
163.099
163.10
≥98.0% (GC,T)
122°C
170886-13-2
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-(trifluoromethyl)-1H-pyridin-4-one
10953849
Crystal-Powder at 20°C
4-Hydroxy-2-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

TSCA : No