missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-2-(trifluoromethyl)pyridine, 97%

$27.30 - $320.76

Chemical Identifiers

CAS 170886-13-2
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.099
MDL Number MFCD01571202
InChI Key LKHCWMQMRXAPHX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
PubChem CID 10953849
IUPAC Name 2-(trifluoromethyl)-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64380MD View Documents Alfa Aesar™
H64380MD
250mg Each for $27.30
 
AAH6438003 View Documents Alfa Aesar™
H6438003
1g Each for $80.34
 
AAH6438006 View Documents Alfa Aesar™
H6438006
5g Each for $320.76
 
Specifications

Chemical Identifiers

170886-13-2
163.099
LKHCWMQMRXAPHX-UHFFFAOYSA-N
10953849
C1=CNC(=CC1=O)C(F)(F)F
C6H4F3NO
MFCD01571202
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
2-(trifluoromethyl)-1H-pyridin-4-one

Specifications

White
170886-13-2
MFCD01571202
LKHCWMQMRXAPHX-UHFFFAOYSA-N
2-(trifluoromethyl)-1H-pyridin-4-one
10953849
Powder
4-Hydroxy-2-(trifluoromethyl)pyridine
250mg
C6H4F3NO
2-trifluoromethyl pyridin-4-ol, 4-hydroxy-2-trifluoromethyl pyridine, 4-hydroxy-2-trifluoromethylpyridine, 2-trifluoromethyl-4-hydroxypyridine, 2-trifluoromethyl-4-pyridinol, 4-pyridinol, 2-trifluoromethyl, 2-trifluoromethyl-1h-pyridin-4-one, 2-trifluoromethylpyridin-4-ol, 2-trifluoromethyl-1,4-dihydropyridin-4-one, pubchem17703
C1=CNC(=CC1=O)C(F)(F)F
163.099
163.1
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning