Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-2-(trifluoromethyl)benzoic acid, 98%

$73.53 - $73.53

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)benzoic acid
CAS 320-32-1
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD01091007
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26127MD

  SDS
alfa aesar™
H26127MD
250mg Each for $73.53
Description & Specifications

Specifications

Formula Weight 206.12
Melting Point 159° to 161°C
Quantity 250mg
Assay 98%
Health Hazard 1 Warning
CAS 320-32-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD01091007
Molecular Formula C8H5F3O3
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)benzoic acid

Applications
4-Hydroxy-2-(trifluoromethyl)benzoic acid is used in pharm industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.