Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester, 97%

$89.04 - $265.19

Chemical Identifiers

Chemical Name or Material 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester
CAS 1073371-73-9
Molecular Formula C16H20BNO3S
Formula Weight 317.21
MDL Number MFCD09953504
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-alpha-(2-thiazolyl)benzyl alcohol; [alpha-(2-Thiazolyl)-4-(hydroxymethyl)]benzeneboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2879803

  SDS
alfa aesar™
H2879803
1g Each for $89.04

AAH2879806

  SDS
alfa aesar™
H2879806
5g Each for $265.19
Description & Specifications

Specifications

Formula Weight 317.21
Chemical Name or Material 4-[Hydroxy(2-thiazolyl)methyl]benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C16H20BNO3S
MDL Number MFCD09953504
CAS 1073371-73-9
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-alpha-(2-thiazolyl)benzyl alcohol; [alpha-(2-Thiazolyl)-4-(hydroxymethyl)]benzeneboronic acid pinacol ester
Melting Point 147° to 150°C
Quantity 1g