missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-2-methylbenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53068MD

 View more versions of this product

Catalog No. AAH53068MD



Specifications

Specifications

4-Hydroxy-2-methylbenzeneboronic acid
250mg
C7H9BO3
4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
B(C1=C(C=C(C=C1)O)C)(O)O
151.956
151.96
100°C to 104°C
493035-82-8
MFCD03788424
OYIYNIONWDBJIF-UHFFFAOYSA-N
(4-hydroxy-2-methylphenyl)boronic acid
2773448
98%

Chemical Identifiers

493035-82-8
151.956
OYIYNIONWDBJIF-UHFFFAOYSA-N
2773448
B(C1=C(C=C(C=C1)O)C)(O)O
C7H9BO3
MFCD03788424
4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
(4-hydroxy-2-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents