Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide, 97%

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide
CAS 59804-37-4
Molecular Formula C13H11N3O4S2
Formula Weight 337.37
MDL Number MFCD00083502
UN Number UN2811
Synonym Tenoxicam
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6083714

  SDS
alfa aesar™
H6083714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.37
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H11N3O4S2
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Chemical Name or Material 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide
UN Number UN2811
Quantity 25g
Health Hazard 1 Danger
CAS 59804-37-4
Assay 97%
MDL Number MFCD00083502
Synonym Tenoxicam
RTECS Number XJ9095060
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.