missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, TCI America™
$103.89 - $362.09
Chemical Identifiers
CAS | 2836-44-4 |
---|---|
Molecular Formula | C6H5F3N2O |
Molecular Weight (g/mol) | 178.114 |
MDL Number | MFCD03265321 |
InChI Key | DBDRDWKBAGNHHS-UHFFFAOYSA-N |
Synonym | 2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl |
PubChem CID | 233901 |
IUPAC Name | 2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one |
SMILES | CC1=NC(=O)C=C(N1)C(F)(F)F |
Chemical Identifiers
2836-44-4 | |
178.114 | |
DBDRDWKBAGNHHS-UHFFFAOYSA-N | |
233901 | |
CC1=NC(=O)C=C(N1)C(F)(F)F |
C6H5F3N2O | |
MFCD03265321 | |
2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl | |
2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one |
Specifications
White-Yellow | |
1g | |
C6H5F3N2O | |
2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl | |
CC1=NC(=O)C=C(N1)C(F)(F)F | |
178.114 | |
178.11 | |
≥98.0% (GC) |
143°C | |
2836-44-4 | |
MFCD03265321 | |
DBDRDWKBAGNHHS-UHFFFAOYSA-N | |
2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one | |
233901 | |
Crystal-Powder at 20°C | |
4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine |
Safety and Handling
TSCA : No