missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-2(5H)-furanone, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 235260010

 View more versions of this product

Catalog No. AC235260010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Hydroxy-2(5H)-furanone
94.0
135°C to 137°C
Authentic
Glass bottle
MFCD00052187
4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g
JZQBAGOECGRTSA-UHFFFAOYSA-N
3-hydroxy-2H-furan-5-one
54683813
95%
541-57-1
100.0
Beige-Yellow
94% min. (HPLC)
C4H4O3
1 g
Solubility in water: soluble. Other solubilities: soluble in alcohol,acetonitrile,dmso
OC1=CC(=O)OC1
100.07
100.07
Crystalline Powder

Chemical Identifiers

541-57-1
100.07
JZQBAGOECGRTSA-UHFFFAOYSA-N
54683813
OC1=CC(=O)OC1
C4H4O3
MFCD00052187
4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g
3-hydroxy-2H-furan-5-one
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 208-785-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only