Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Hydroxy-2,4-dimethoxybenzophenone, 98%

Chemical Identifiers

Chemical Name or Material 4'-Hydroxy-2,4-dimethoxybenzophenone
CAS 41351-30-8
Formula Weight 258.27
MDL Number MFCD00137850
Beilstein 2508320
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6621714

  SDS
alfa aesar™
H6621714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6621706

  SDS
alfa aesar™
H6621706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6621722

  SDS
alfa aesar™
H6621722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.27
Melting Point 138° to 142°C
Chemical Name or Material 4'-Hydroxy-2,4-dimethoxybenzophenone
Physical Form Powder
Assay 98%
MDL Number MFCD00137850
CAS 41351-30-8
Beilstein 2508320
Storage Note 1 2° to 8°C
Color Off-white to white
Quantity 25g
Safety and Handling

2° to 8°C