Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-1H-benzotriazole, 98%

$82.12 - $188.35

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-1H-benzotriazole
CAS 26725-51-9
Molecular Formula C6H5N3O
Formula Weight 135.13
MDL Number MFCD00970892
Synonym 1H-Benzotriazol-4-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2584303

  SDS
alfa aesar™
H2584303
1g Each for $188.35

AAH25843MD

  SDS
alfa aesar™
H25843MD
250mg Each for $82.12
Description & Specifications

Specifications

Formula Weight 135.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 1H-Benzotriazol-4-ol
Chemical Name or Material 4-Hydroxy-1H-benzotriazole
Melting Point 221° to 223°C
Health Hazard 1 Warning
Assay 98%
CAS 26725-51-9
Molecular Formula C6H5N3O
MDL Number MFCD00970892
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.