Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine, 98+%

$500.27 - $500.27

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
CAS 21314-17-0
Molecular Formula C11H8N4O
Formula Weight 212.21
MDL Number MFCD00082637
Beilstein 198880
Synonym 1,5-Dihydro-1-phenyl-4H-pyrazolo[3,4-d]pyrimidin-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1028606

 
alfa aesar™
L1028606
5g Each for $500.27
Description & Specifications

Specifications

Formula Weight 212.21
Chemical Name or Material 4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
MDL Number MFCD00082637
Beilstein 198880
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Molecular Formula C11H8N4O
CAS 21314-17-0
Synonym 1,5-Dihydro-1-phenyl-4H-pyrazolo[3,4-d]pyrimidin-4-one
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 304° to 307°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.