missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar L1028603

 View more versions of this product

Catalog No. AAL1028603



Specifications

Specifications

4-Hydroxy-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
1g
C11H8N4O
198880
SVKPVSOBZKPDIY-UHFFFAOYSA-N
1-phenyl-2H-pyrazolo[3,4-d]pyrimidin-4-one
88867
≥98%
304°C to 307°C
21314-17-0
MFCD00082637
1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ol, 4-hydroxy-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one, 4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5-dihydro-1-phenyl, 1-phenyl-2h-pyrazolo 3,4-d pyrimidin-4-one, gdzqghgckkwnac-uhfffaoysa-n, 1-phenylpyrazolo 3,4-d pyrimidin-4-ol, 1-phenyl-5h-pyrazolo 3,4-d pyrimidin-4-one, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4 5h-one, 1-phenyl-5-hydropyrazolo 5,4-d pyrimidin-4-one
C1=CC=C(C=C1)N2C3=NC=NC(=O)C3=CN2
212.212
212.21

Chemical Identifiers

21314-17-0
212.212
SVKPVSOBZKPDIY-UHFFFAOYSA-N
88867
C1=CC=C(C=C1)N2C3=NC=NC(=O)C3=CN2
C11H8N4O
MFCD00082637
1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4-ol, 4-hydroxy-1-phenylpyrazolo 3,4-d pyrimidine, 1-phenyl-1,5-dihydro-4h-pyrazolo 3,4-d pyrimidin-4-one, 4h-pyrazolo 3,4-d pyrimidin-4-one, 1,5-dihydro-1-phenyl, 1-phenyl-2h-pyrazolo 3,4-d pyrimidin-4-one, gdzqghgckkwnac-uhfffaoysa-n, 1-phenylpyrazolo 3,4-d pyrimidin-4-ol, 1-phenyl-5h-pyrazolo 3,4-d pyrimidin-4-one, 1-phenyl-1h-pyrazolo 3,4-d pyrimidin-4 5h-one, 1-phenyl-5-hydropyrazolo 5,4-d pyrimidin-4-one
1-phenyl-2H-pyrazolo[3,4-d]pyrimidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents