missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-1-indanone, TCI America™

$279.59 - $948.77

Chemical Identifiers

CAS 40731-98-4
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00143330
InChI Key CKSCMRNFDBWFND-UHFFFAOYSA-N
Synonym 4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
PubChem CID 590547
IUPAC Name 4-hydroxy-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C(=CC=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H07235G View Documents TCI America
H07235G
5g Each for $279.59
 
H072325G View Documents TCI America
H072325G
25g Each for $948.77
 
Specifications

Chemical Identifiers

40731-98-4
148.161
CKSCMRNFDBWFND-UHFFFAOYSA-N
590547
C1CC(=O)C2=C1C(=CC=C2)O
C9H8O2
MFCD00143330
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
4-hydroxy-2,3-dihydroinden-1-one

Specifications

White-Yellow
40731-98-4
MFCD00143330
CKSCMRNFDBWFND-UHFFFAOYSA-N
4-hydroxy-2,3-dihydroinden-1-one
590547
Crystal-Powder at 20°C
4-Hydroxy-1-indanone
5g
C9H8O2
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659
C1CC(=O)C2=C1C(=CC=C2)O
148.161
148.16
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No