Please login to your online account to display your discounted pricing

4-Hydroxy-1-indanone, 97%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC399030050

 View more versions of this product

Catalog No. AC399030050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-1-indanone
Quantity 5g
CAS 40731-98-4
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H8O2
Formula Weight 148.16

Specifications

Chemical Name or Material 4-Hydroxy-1-indanone
Assay Percent Range 97%
Molecular Formula C9H8O2
Formula Weight 148.16