Please login to your online account to display your discounted pricing

4-Hydroxy-1-indanone, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Hydroxy-1-indanone
Assay Percent Range 97%
Molecular Formula C9H8O2
Formula Weight 148.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399030010

  SDS
thermo fisher scientific
AC399030010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399030050

 
thermo fisher scientific
AC399030050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.16
Quantity 1g
Chemical Name or Material 4-Hydroxy-1-indanone
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H8O2
CAS 40731-98-4