missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Hydroxy-1-indanone, 97%

Manufacturer:  Alfa Aesar™ H5618403

 View more versions of this product

Catalog No. AAH5618403



Description

Description

Applications
4-Hydroxy-1-indanone is a hydroxylated indanone used for synthesis. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in ethanol. (25mg/ml)

Notes
Incompatible with oxidizing agents. Store at room temperature. Avoid excessive heat and light.
Specifications

Specifications

4-Hydroxy-1-indanone
1g
C9H8O2
1h-inden-1-one, 2,3-dihydro-4-hydroxy
CKSCMRNFDBWFND-UHFFFAOYSA-N
4-hydroxy-2,3-dihydroinden-1-one
590547
97%
242°C to 244°C
40731-98-4
MFCD00143330
Soluble in ethanol. (25mg/ml)
C1CC(=O)C2=C1C(=CC=C2)O
148.161
148.16

Chemical Identifiers

40731-98-4
148.161
CKSCMRNFDBWFND-UHFFFAOYSA-N
590547
C1CC(=O)C2=C1C(=CC=C2)O
C9H8O2
MFCD00143330
1h-inden-1-one, 2,3-dihydro-4-hydroxy
4-hydroxy-2,3-dihydroinden-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents