missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydrazinobenzoic acid hydrochloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 24589-77-3 |
---|---|
Molecular Formula | C7H9ClN2O2 |
Molecular Weight (g/mol) | 188.61 |
MDL Number | MFCD00039073 |
InChI Key | XHLQMKQBCHYRLC-UHFFFAOYSA-N |
Synonym | 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride |
PubChem CID | 90547 |
IUPAC Name | 4-hydrazinylbenzoic acid;hydrochloride |
SMILES | [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC450670100
|
Thermo Scientific Chemicals
450670100 |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC450670500
|
Thermo Scientific Chemicals
450670500 |
50g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC450672500
|
Thermo Scientific Chemicals
450672500 |
250g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
24589-77-3 | |
188.61 | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
90547 | |
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O |
C7H9ClN2O2 | |
MFCD00039073 | |
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride | |
4-hydrazinylbenzoic acid;hydrochloride |
Specifications
24589-77-3 | |
Glass bottle | |
MFCD00039073 | |
4-hydrazinobenzoic acid hydrochloride, 4-hydrazinylbenzoic acid hydrochloride, p-hydrazinobenzoic acid hydrochloride, 4-carboxyphenylhydrazine hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, 4-carboxyphenylhydrazinium chloride, p-carboxyphenylhydrazine hydrochloride, ccris 7999, p-hydrazinobenzoic acid.hcl, 4-hydrazinobenzoic acid monohydrochloride | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
4-hydrazinylbenzoic acid;hydrochloride | |
90547 | |
97% |
96% min. (Argentometry) | |
C7H9ClN2O2 | |
10g | |
Solubility in water: soluble. | |
[H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O | |
188.61 | |
188.61 | |
4-Hydrazinobenzoic acid hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 246-330-
RUO â Research Use Only