missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydrazinobenzoic acid hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450670500

 View more versions of this product

Catalog No. AC450670500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Hydrazinobenzoic acid hydrochloride
252.0°C to 254.0°C
24589-77-3
C7H8N2O2·HCl
Solubility in water: soluble.
C1=CC(=CC=C1C(=O)O)NN.Cl
188.61
188.61
Glass bottle
50g
96% min. (Argentometry)
4-carboxyphenylhydrazine hydrochloride, 4-carboxyphenylhydrazinium chloride, 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinobenzoic acid monohydrochloride, 4-hydrazinylbenzoic acid hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, ccris 7999, p-carboxyphenylhydrazine hydrochloride, p-hydrazinobenzoic acid hydrochloride, p-hydrazinobenzoic acid.hcl
XHLQMKQBCHYRLC-UHFFFAOYSA-N
4-hydrazinylbenzoic acid;hydrochloride
90547
97%

Chemical Identifiers

24589-77-3
188.61
4-carboxyphenylhydrazine hydrochloride, 4-carboxyphenylhydrazinium chloride, 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinobenzoic acid monohydrochloride, 4-hydrazinylbenzoic acid hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, ccris 7999, p-carboxyphenylhydrazine hydrochloride, p-hydrazinobenzoic acid hydrochloride, p-hydrazinobenzoic acid.hcl
4-hydrazinylbenzoic acid;hydrochloride
C7H8N2O2·HCl
XHLQMKQBCHYRLC-UHFFFAOYSA-N
90547
C1=CC(=CC=C1C(=O)O)NN.Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 246-330-

SDS
Documents