missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Hydrazinobenzoic acid hydrochloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450670100
Specifications
4-Hydrazinobenzoic acid hydrochloride | |
252.0°C to 254.0°C | |
24589-77-3 | |
C7H8N2O2·HCl | |
Solubility in water: soluble. | |
C1=CC(=CC=C1C(=O)O)NN.Cl | |
188.61 | |
188.61 |
Glass bottle | |
10g | |
96% min. (Argentometry) | |
4-carboxyphenylhydrazine hydrochloride, 4-carboxyphenylhydrazinium chloride, 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinobenzoic acid monohydrochloride, 4-hydrazinylbenzoic acid hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, ccris 7999, p-carboxyphenylhydrazine hydrochloride, p-hydrazinobenzoic acid hydrochloride, p-hydrazinobenzoic acid.hcl | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
4-hydrazinylbenzoic acid;hydrochloride | |
90547 | |
97% |
Chemical Identifiers
24589-77-3 | |
188.61 | |
4-carboxyphenylhydrazine hydrochloride, 4-carboxyphenylhydrazinium chloride, 4-hydrazinobenzoic acid hydrochloride, 4-hydrazinobenzoic acid monohydrochloride, 4-hydrazinylbenzoic acid hydrochloride, benzoic acid, 4-hydrazino-, monohydrochloride, ccris 7999, p-carboxyphenylhydrazine hydrochloride, p-hydrazinobenzoic acid hydrochloride, p-hydrazinobenzoic acid.hcl | |
4-hydrazinylbenzoic acid;hydrochloride |
C7H8N2O2·HCl | |
XHLQMKQBCHYRLC-UHFFFAOYSA-N | |
90547 | |
C1=CC(=CC=C1C(=O)O)NN.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 246-330-