Please login to your online account to display your discounted pricing

4-Hydrazinobenzoic acid, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC411795000

 View more versions of this product

Catalog No. AC411795000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 152.15
Quantity 500g
Color Light yellow to light brown
CAS Max % 100.0
MDL Number MFCD00007581
CAS 619-67-0
CAS Min % 97.5
Health Hazard 1 Warning
Chemical Name or Material 4-Hydrazinobenzoic acid
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C7H8N2O2
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Hydrazinobenzoic acid
CAS 619-67-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD00007581
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents