Please login to your online account to display your discounted pricing

4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  411781000

 View more versions of this product

Catalog No. AC411781000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 197
Quantity 100g
Color Light yellow to slightly orange
Merck Index 14, 4773
EINECS Number 202-694-2
TSCA TSCA
RTECS Number DB6900000
CAS Max % 100.0
Infrared Spectrum Authentic
Loss on Drying 4% to 5%
Packaging Glass bottle
Synonym Phenylhydrazine-4-sulfonic acid, hemihydrate
CAS Min % 97.5
CAS 98-71-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Hydrazinobenzenesulfonic acid, hemihydrate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H8N2O3S·1/2H2O
MDL Number MFCD00480958
CAS 854689-07-9
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-Hydrazinobenzenesulfonic acid, hemihydrate
Name Note 98%
CAS 854689-07-9
CAS Min % 97.5
CAS Max % 100.0
CAS 98-71-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H8N2O3S·1/2H2O
Formula Weight 197
MDL Number MFCD00480958
Synonym Phenylhydrazine-4-sulfonic acid, hemihydrate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents