Please login to your online account to display your discounted pricing

4-Hydrazinobenzene-1-sulfonamide hydrochloride, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Hydrazinobenzene-1-sulfonamide hydrochloride
CAS 17852-52-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H9N3O2S·ClH
Linear Formula NH2NHC6H4SO2NH2·HCl
Formula Weight 223.68
MDL Number MFCD00052262
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB06645EA

 
fisher scientific company
BTB06645EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB06645FA

 
fisher scientific company
BTB06645FA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB06645EE

 
fisher scientific company
BTB06645EE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.68
Quantity 10g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-Hydrazinobenzene-1-sulfonamide hydrochloride
Assay Percent Range 95%
Molecular Formula C6H9N3O2S·ClH
Linear Formula NH2NHC6H4SO2NH2·HCl
Packaging Amber Glass Bottle
MDL Number MFCD00052262
CAS 17852-52-7
CAS Min % 94.5